Order Entry
Switzerland
ContactUsLinkComponent
Antistatic mats, b.safe
Antistatic mats, b.safe
Codice catalogo 590-0346
Nome del fornitore:  Bohlender
undefined
Antistatic mats, b.safe
Codice catalogo 590-0346
Nome del fornitore:  Bohlender

Specifiche

  • Description:
    Antistatic mat, with 1,5 m grounding cable with clip
  • L×W:
    1200×600 mm
  • Pk:
    1

Specifiche

Informazioni su questo articolo

Mat made of static dissipative material with permanently mounted grounding cable including clip for earthing connection.

  • Suitable grounding accessories for b.safe products
  • Customised versions can be offered on request

For grounding vessels made of static dissipative materials or persons before handling with easy flammable liquids.