ContactUsLinkComponent
Face Shields, V5 series
Face Shields, V5 series
Catalog # MMMA5F-11
Supplier:  3M
undefined
Face Shields, V5 series
Catalog # MMMA5F-11
Supplier:  3M
Specifications
  • Colour:
    Transparent
  • Description:
    V5 series faceshield
  • L×W:
    244×462 mm
  • Pk:
    1
Specifications
About this item
Optical class 1, clear curved polycarbonate faceshield with anti-fog and anti-scratch coating. Compatible with 3M™ V5 Faceshield System for mounting on 3M™ Hard Hat Series G3000 or G22.
  • Provides protection against secondary welding light
  • Anti-fog and anti-scratch coated
  • Snap in feature makes it quick and easy to connect
  • Medium energy impact, Grade B
  • Optical class 1 visor, low energy impact
Get protection against impacts and secondary welding light with 3M™ Faceshield, 5F-11. It provides impact protection B against medium speed particles (0.86 g steel ball at 120 m/s), UV, or intense visible light. Anti-fog and anti-scratch coated, with a curved shape that allows for a close fit to the face.
Designed to fit 3M™ Hard Hat Series G3000 or G22 with 3M™ V5 Faceshield Mounting system. The snap in feature makes it quick and easy for the faceshield to connect to the Faceshield Holder.
Certifications: EN 166:2001