Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant CD200 R1 (from HEK293)
Anbieter:  ACRO Biosystems
Zertifikate
Human recombinant CD200 R1 (from HEK293)
Anbieter:  ACRO Biosystems

Technische Daten

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human CD200, Fc Tag at 2 μg/ml (100 μl/well) can bind Biotinylated Human CD200 R1, Fc,Avitag with a linear range of 0,5-7,8 ng/ml (QC tested)
  • Gene ID:
    Q8TD46-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    MOX2R|CD200R|OX2R|CRTR2
  • Protein/peptide name:
    CD200 R1
  • Purity:
    >95%
  • Molecular weight:
    55,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Technische Daten

Frequently Bought Together