Order Entry
Switzerland
ContactUsLinkComponent
 
Human recombinant B7-H6 (from HEK293)
 
Human recombinant B7-H6 (from HEK293)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human NKp30, Fc Tag at 10 μg/ml (100 μl/well) can bind Human B7-H6, His Tag with a linear range of 0,016-0,25 μg/ml (QC tested)
  • Gene ID:
    Q68D85-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    B7-H6|B7 Homolog 6|NCR3LG1
  • Protein/peptide name:
    B7-H6
  • Purity:
    >90%
  • Molecular weight:
    28,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

 

Frequently Bought Together