Rat recombinant IGF-I R (from HEK293)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Rat
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. immobilised Rat IGF-I R, His Tag (Cat. No. ACRBIGR-R5224) at 5 μg/ml (100 μl/well) can bind Human IGF-I, Fc Tag (Cat. No. ACRBIG1-H4269) with a linear range of 0,01 - 0,313 μg/ml (QC tested)
- Gene ID:P24062-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:MGC18216|JTK13|CD221|IGFR|MGC142170|IGF1R|MGC142172
- Protein/peptide name:IGF-I R
- Purity:>90%
- Molecular weight:81,3 kDa and 24,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder, RT
Frequently Bought Together