Mouse recombinant SLAMF7 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Gene ID:Q8BHK6-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:FOAP-12|CRACC|19A|CS1|CD319|SLAMF7
- Protein/peptide name:SLAMF7
- Purity:>95%
- Molecular weight:49,4 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder, RT
Specifications
Frequently Bought Together