Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant BCMA (from HEK293)
Mouse recombinant BCMA (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Mouse recombinant BCMA (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. immobilised Mouse BAFF, Mouse IgG2a Fc Tag at 5 μg/ml (100 μl/well) can bind Mouse BCMA, Fc Tag (Cat. No. ACRBBCA-M5258) with a linear range of 2 - 20 ng/ml (QC tested)
  • Gene ID:
    O88472-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CD269|TNFRSF17|BCM
  • Protein/peptide name:
    BCMA
  • Purity:
    >95%
  • Molecular weight:
    32,1 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together