Human recombinant UCH-L3 (from E. coli)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Gene ID:P15374-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:Ubiquitin thioesterase L3|UCHL3
- Protein/peptide name:UCH-L3
- Purity:>95%
- Molecular weight:27,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder, RT
Frequently Bought Together