Human recombinant CD40 ligand (from HEK293 cells)
Référence ACRBCDL-H52DB-1MG
Fournisseur: ACRO Biosystems
Numéro CAS:
Spécifications
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His, Flag Tag (active trimer) (MALS verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Loaded Human CD40, Fc Tag (HPLC-verified) (Cat. No. CD0-H5253) on Protein A Biosensor, can bind Human CD40 Ligand, His,Flag Tag (active trimer) (MALS verified) (Cat. No. CDL-H52Db) with an affinity constant of 1,58 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Human CD40 Ligand, His,Flag Tag (active trimer) (MALS verified) (Cat. No. CDL-H52Db) stimulates secretion of IL-6 by human B cells. The ED50 for this effect is 18,76 - 19,45 ng/ml in the presence of 5 ng/ml of Recombinant Human IL‑4 (Routinely tested).
- Gene ID:P29965-1
- Protein synonyms:TRAP|CD40L|IMD3|gp39|CD154|CD40LG|IGM|T-BAM|TNFSF5|HIGM1
- Protein/peptide name:CD40 Ligand
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:49,9 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
- Shipping temperature:RT
Spécifications
À propos de cet article
Human CD40 Ligand / TNFSF5 protein, His,Flag Tag (active trimer) (MALS verified).