Order Entry
Switzerland
ContactUsLinkComponent
Cynomolgus monkey, rhesus macaque recombinant BCMA (from HEK293 cells), Biotin
Cynomolgus monkey, rhesus macaque recombinant BCMA (from HEK293 cells), Biotin
Supplier:  ACRO Biosystems
Certificates
Cynomolgus monkey, rhesus macaque recombinant BCMA (from HEK293 cells), Biotin
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
    Rhesus monkey
  • Tag sequence:
    His, Avitag™
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Gene ID:
    G7Q0I4-1
  • Protein synonyms:
    CD269|TNFRSF17|BCM
  • Protein/peptide name:
    BCMA
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    9,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together