Human recombinant BAFF (from HEK293 cells)
Human BAFF / TNFSF13B / CD257 protein, Fc Tag, active trimer (MALS verified).
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Immobilised Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) on CM5 Chip can bind Human TACI/TNFRSF13B, Fc Tag (Cat. No. TAI-H5256) with an affinity constant of 0.658 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Immobilised Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human BAFFR, Llama IgG2b Fc Tag, low endotoxin (Cat. No. BAR-H5258) with an affinity constant of 44,5 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) on Protein A Biosensor, can bind Biotinylated Human BCMA, His,Avitag (Cat. No. BCA-H82E4) with an affinity constant of 0,69 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) stimulates proliferation of mouse spleen cells. The EC50 for this effect is 4,91 - 9,81 ng/ml (Routinely tested).
- Gene ID:Q9Y275-1
- Protein synonyms:CD257|ZTNF4|THANK|TALL-1|TNFSF20|DTL|TNFSF13B|TALL1|BLYS|BAFF
- Protein/peptide name:BAFF
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:76,7 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH 7.5. Normally trehalose is added as protectant before Lyophilisation.
- Shipping temperature:RT
Frequently Bought Together