Human recombinant Alpha-Synuclein (from E. coli)
Marketsource
Human Alpha-Synuclein protein, His Tag.
Human Alpha-Synuclein protein, His Tag.
- Catalog No:
- ACRBALN-H52H8200UG
- ACRBALN-H52H8-1MG
- Protein/peptide name:
- Alpha-Synuclein
- Alpha-Synuclein
- Protein synonyms:
- PARK1|SNCA|alpha-Synuclein|NACP
- PARK1|SNCA|alpha-Synuclein|NACP
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Source:
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Gene ID:
- P37840-1
- P37840-1
- Endotoxin level:
- Low
- Low
- Endotoxin content:
- Less than 1,0 EU per μg by the LAL method.
- Less than 1,0 EU per μg by the LAL method.
- Purity:
- >90% as determined by SDS-PAGE.
- >90% as determined by SDS-PAGE.
- Formulation:
- Lyophilised from 0,22 μm filtered solution in 100 mM NaAC, pH 7,0. Normally trehalose is added as protectant before Lyophilisation.
- Lyophilised from 0,22 μm filtered solution in 100 mM NaAC, pH 7,0. Normally trehalose is added as protectant before Lyophilisation.
- Molecular weight:
- 16,3 kDa
- 16,3 kDa
- Storage conditions:
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- Tag sequence:
- His Tag
- His Tag
Product Family Options
- Pk
- Marketsource
Specifications
Cat. No.ACRBALN-H52H8200UGProtein/peptide nameAlpha-SynucleinProtein synonymsPARK1|SNCA|alpha-Synuclein|NACPProtein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP37840-1Endotoxin levelLowEndotoxin contentLess than 1,0 EU per μg by the LAL method.Purity>90% as determined by SDS-PAGE.FormulationLyophilised from 0,22 μm filtered solution in 100 mM NaAC, pH 7,0. Normally trehalose is added as protectant before Lyophilisation.Molecular weight16,3 kDaStorage conditions–20 °CShipping temperatureRTTag sequenceHis TagMarketsource - Marketsource
Specifications
Cat. No.ACRBALN-H52H8-1MGProtein/peptide nameAlpha-SynucleinProtein synonymsPARK1|SNCA|alpha-Synuclein|NACPProtein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP37840-1Endotoxin levelLowEndotoxin contentLess than 1,0 EU per μg by the LAL method.Purity>90% as determined by SDS-PAGE.FormulationLyophilised from 0,22 μm filtered solution in 100 mM NaAC, pH 7,0. Normally trehalose is added as protectant before Lyophilisation.Molecular weight16,3 kDaStorage conditions–20 °CShipping temperatureRTTag sequenceHis TagMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



