Rhesus macaque recombinant LAIR-1 (from HEK293 cells)
Katalognr. ACRBLA1-C52A3-1MG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Tag sequence:Mouse IgG2a Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised Human Collagen I protein at 2 μg/ml (100 μl/well) can bind Rhesus macaque LAIR-1, Mouse IgG2a Fc Tag (Cat. No. LA1-C52A3) with a linear range of 5 - 78 ng/ml (QC tested).
- Gene ID:ALV66454.1
- Protein synonyms:LAIR1|CD305
- Protein/peptide name:LAIR-1
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:42,9 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before Lyophilisation.
- Shipping temperature:RT
Specifications
About this item
Rhesus macaque LAIR-1 / CD305 protein, Mouse IgG2a Fc Tag.