Rabbit recombinant Complement C5 (from HEK293 cells)
Marketsource
Supplier: ACRO Biosystems
Rabbit recombinant Complement C5 (from HEK293 cells)
Supplier: ACRO Biosystems
Marketsource
Certificates
About this item
Rabbit Complement C5 protein, His Tag.
2 Options Available Below
Product Details & Documents
Rabbit Complement C5 protein, His Tag.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ACRBCO5-R52H5100UG
- ACRBCO5-R52H5-1MG
- Protein/peptide name:
- Complement C5
- Complement C5
- Protein synonyms:
- CPAMD4|Complement C5|C5
- CPAMD4|Complement C5|C5
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Rabbit
- Rabbit
- Source:
- HEK293 cells
- HEK293 cells
- Conjugation:
- Unconjugated
- Unconjugated
- Endotoxin level:
- Low
- Low
- Endotoxin content:
- Less than 1,0 EU per μg by the LAL method.
- Less than 1,0 EU per μg by the LAL method.
- Purity:
- >90% as determined by SDS-PAGE.
- >90% as determined by SDS-PAGE.
- Formulation:
- Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
- Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
- Molecular weight:
- 73,8 kDa (β chain) and 111,9 kDa (α chain)
- 73,8 kDa (β chain) and 111,9 kDa (α chain)
- Storage conditions:
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- Tag sequence:
- His Tag
- His Tag
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.ACRBCO5-R52H5100UGProtein/peptide nameComplement C5Protein synonymsCPAMD4|Complement C5|C5Protein/peptide typeRecombinantSpeciesRabbitSourceHEK293 cellsConjugationUnconjugatedEndotoxin levelLowEndotoxin contentLess than 1,0 EU per μg by the LAL method.Purity>90% as determined by SDS-PAGE.FormulationLyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.Molecular weight73,8 kDa (β chain) and 111,9 kDa (α chain)Storage conditions–20 °CShipping temperatureRTTag sequenceHis TagMarketsource - Specifications:Marketsource
More
Specifications
Cat. No.ACRBCO5-R52H5-1MGProtein/peptide nameComplement C5Protein synonymsCPAMD4|Complement C5|C5Protein/peptide typeRecombinantSpeciesRabbitSourceHEK293 cellsConjugationUnconjugatedEndotoxin levelLowEndotoxin contentLess than 1,0 EU per μg by the LAL method.Purity>90% as determined by SDS-PAGE.FormulationLyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.Molecular weight73,8 kDa (β chain) and 111,9 kDa (α chain)Storage conditions–20 °CShipping temperatureRTTag sequenceHis TagMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



