Human recombinant sonic hedgehog (from HEK293)
Catalog # ACRBSH7-H5229-1MG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyopilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Gene ID:Q15465
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:TPT|Sonic hedgehog|SHH|HLP3|SMMCI|HPE3|TPTPS|HHG-1
- Protein/peptide name:Sonic Hedgehog
- Purity:>95%
- Molecular weight:20,4 kDa
- Endotoxin level:Low
- Formulation:Lyopilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT