Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant IL-3 R alpha (from HEK293 cells)
Mouse recombinant IL-3 R alpha (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Mouse recombinant IL-3 R alpha (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Tag sequence:
    His Tag (SPR verified)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Gene ID:
    P26952-1
  • Protein synonyms:
    IL3RX|IL3RA|CD123 antigen|IL-3Ra|IL3RY|CD123|IL-3 R alpha|hIL3Ra|MGC34174|hIL-3Ra|IL3RAY|IL3R|IL-3R-alpha
  • Protein/peptide name:
    IL-3 R alpha
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    36,4 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together