Cynomolgus recombinant SIRP alpha (from HEK293)
Catalog # ACRBSIA-C52H7-1MG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pack type:1 mg
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Cynomolgus / Rhesus macaque CD47, Fc Tag at 2 μg/ml (100 μl/well) can bind Cynomolgus SIRP alpha, His Tag with a linear range of 0,313-2,5 μg/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Biotinylated Human CD47, Fc,Avitag at 1 μg/ml (100 μl/well) on streptavidin precoated (0,2 ug/well) plate, can bind Cynomolgus SIRP alpha, His Tag with a linear range of 0,039-0,625 μg/ml (Routinely tested)
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:MYD1|PTPNS1|CD172A|SHPS1|P84|SIRPA|BIT|MFR
- Protein/peptide name:SIRP alpha
- Purity:>95%
- Molecular weight:39,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder,RT