Mouse recombinant IFN-alpha 1 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Mouse IFN-alpha 1 Protein, His Tag.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Gene ID:P01572-1
- Protein synonyms:LeIF D|IFNA1|IFN-alpha-1|IFN-alpha 1|IFNalpha 1
- Protein/peptide name:IFN-alpha 1
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:21,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Shipping temperature:RT
Specifications
Frequently Bought Together