Mouse recombinant Fc gamma RIV/ CD16-2 (from HEK293 cells)
Mouse Fc gamma RIV / CD16-2 Protein, His Tag.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Captured Mouse CD16-2, His Tag (ACRBFC4-M52H3) on NTA-Series S sensor chip can bind OKT3 with an affinity constant of 154 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Mouse CD16-2, His Tag (ACRBFC4-M52H3) on NTA Biosensor, can bind OKT3 with an affinity constant of 140 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Mouse CD16-2, His Tag (ACRBFC4-M52H3) on NTA Biosensor, can bind Herceptin with an affinity constant of 122 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:A0A0B4J1G0-1
- Protein synonyms:Fcgr4|Fc gamma RIV|CD16-2
- Protein/peptide name:Fc gamma RIV/ CD16-2
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:22,8 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Shipping temperature:RT
Frequently Bought Together