Human recombinant NKG2C (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:P26717-1
- Protein synonyms:NKG2-C|KLRC2|NK cell receptor C|CD159c|NKG2C
- Protein/peptide name:NKG2C
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:17,8 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together