Human recombinant netrin receptor DCC (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag (MALS verified)
- Storage conditions:–20 °C
- Gene ID:P43146-1
- Protein synonyms:Netrin receptor DCC|IGDCC1|netrin receptor DCC|DCC
- Protein/peptide name:Netrin receptor DCC
- Molecular weight:119,5 kDa
- Endotoxin level:Low
- Formulation:Please contact us for detailed information.
- Shipping temperature:RT
Specifications
Frequently Bought Together