Human recombinant MIS RII (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:Q16671-1
- Protein synonyms:MISR2|AMHR|AMH type II receptor|MRII|AMHR2|MIS RII
- Protein/peptide name:MIS RII
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:40,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together