Human recombinant LIGHT (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag (active trimer) (MALS verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Human LIGHT, Fc Tag (active trimer) (MALS verified) induced cytotoxicity in HT-29 cells. The ED50 for this effect is 1,29 to 2,38 ng/ml (Routinely tested).
- Gene ID:O43557-1
- Protein synonyms:HVEM-L|CD258|LTg|TR2|LIGHT|HVEML|CD258 antigen|TNFSF14
- Protein/peptide name:Light
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:82,6 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together