Human recombinant IL-7-P2A (from HEK293 cells)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Tag Free
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:P13232-1
- Protein synonyms:Interleukin-7-P2A|IL-7-P2A
- Protein/peptide name:IL-7-P2A
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:19,6 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Frequently Bought Together