Human recombinant IL-2 R gamma (from HEK293 cells)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:P31785
- Protein synonyms:CD132|CIDX|IL2RG|SCIDX1|IMD4|gammaC|P64|SCIDX
- Protein/peptide name:IL-2 R gamma
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:53,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Frequently Bought Together