Mouse recombinant Fc gamma RIII/ CD16 (from HEK293 cells)
Codice catalogo ACRBCDA-M52H8-50UG
Nome del fornitore: ACROBIOSYSTEMS
Numero CAS:
Specifiche
- Pk:50 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag (SPR & BLI verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Mouse CD16, His Tag (SPR & BLI verified) (ACRBCDA-M52H8) Captured on NTA-Series S sensor chip can bind OKT3 with an affinity constant of 0,126 μM as determined in a SPR assay (Biacore T200) (QC tested). Loaded Mouse CD16, His Tag (SPR & BLI verified) (ACRBCDA-M52H8) on NTA Biosensor, can bind OKT3 with an affinity constant of 0,78 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Mouse CD16, His Tag (SPR & BLI verified) (ACRBCDA-M52H8) on NTA Biosensor, can bind Herceptin with an affinity constant of 2,8 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q5D5I8
- Protein synonyms:FCGR3
- Protein/peptide name:Fc gamma RIII/ CD16
- Purity:>95% as determined by reduced SDS-PAGE.
- Molecular weight:23,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Shipping temperature:RT
Specifiche
Informazioni su questo articolo
Mouse Fc gamma RIII / CD16 Protein, His Tag (SPR & BLI verified).