Human recombinant LILRA6 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised human ANGPTL7, His tag (Cat. No. ACRBAN7-H52H5) at 5 μg/ml (100 μl/well) can bind biotinylated human LILRA6, His,Avitag (Cat. No. ACRBLI6-H82E2) with a linear range of 0,078 - 0,625 μg/ml (QC tested).
- Gene ID:Q6PI73-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD85b|ILT8|ILT-8|LILRA6
- Protein/peptide name:LILRA6
- Purity:>95%
- Molecular weight:50,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together