Human recombinant integrin alpha V beta 5 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised vitronectin at 2 μg/ml (100 μl/well) can bind human ITGAV and ITGB5 heterodimer protein, His tag and tag (Cat. No. ACRBIT5-H52W5) with a linear range of 2 - 20 ng/ml (QC tested).
- Gene ID:P06756-1 (V783I) (IL23A) and P18084-1 (B2M)
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:Integrin alpha V beta 5|ITGAV and ITGB5
- Protein/peptide name:Integrin alpha V beta 5
- Purity:>95%
- Molecular weight:112,9 kDa (ITGAV) and 81,9 kDa (ITGB5)
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together