Order Entry
Switzerland
ContactUsLinkComponent
 
Human recombinant integrin alpha E beta 7 (from HEK293)
Catalog # ACRBIT7-H52W7-50UG
Supplier:  ACRO Biosystems
CAS Number:  
Human recombinant integrin alpha E beta 7 (from HEK293)
Catalog # ACRBIT7-H52W7-50UG
Supplier:  ACRO Biosystems
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Gene ID:
    P38570-1 (ITGAE) and P26010-1 (ITGB7)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    Integrin alpha E beta 7|ITGAE and ITGB7
  • Protein/peptide name:
    Integrin alpha E beta 7
  • Purity:
    >90%
  • Molecular weight:
    128,6 kDa (ITGAE) and 81,8 kDa (ITGB7)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

About this item