Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant integrin alpha 4 beta 1 (from HEK293)
Human recombinant integrin alpha 4 beta 1 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant integrin alpha 4 beta 1 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised natalizumab at 1 μg/ml (100 μl/well) can bind biotinylated human ITGA4 and ITGB1 heterodimer protein, His,Avitag and tag free (Cat. No. ACRBIT1-H82W1) with a linear range of 2 - 31 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Human VCAM-1, His Tag (Cat. No. ACRBVC1-H5224) at 5 μg/ml (100 μl/well) can bind biotinylated human ITGA4 and ITGB1 heterodimer protein, His,Avitag and tag free (Cat. No. ACRBIT1-H82W1) with a linear range of 0,01 - 0,156 μg/ml (routinely tested).
  • Gene ID:
    P13612-1 (ITGA4) and P05556-1 (ITGB1)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    Integrin alpha 4 beta 1|ITGA4 and ITGB1
  • Protein/peptide name:
    Integrin alpha 4 beta 1
  • Purity:
    >95%
  • Molecular weight:
    113,2 kDa (ITGA4) and 83,7 kDa (ITGB1)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together