Order Entry
Switzerland
ContactUsLinkComponent
Cynomolgus recombinant LAG-3 (from HEK293)
Cynomolgus recombinant LAG-3 (from HEK293)
Catalog # ACRBLA3-C52A0100UG
Supplier:  ACRO Biosystems
CAS Number:  
Cynomolgus recombinant LAG-3 (from HEK293)
Catalog # ACRBLA3-C52A0100UG
Supplier:  ACRO Biosystems
CAS Number:  

Specifications

  • Pack type:
    100 µg
  • Pk:
    100 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Cynomolgus LAG-3, Mouse IgG2a Fc tag at 2 μg/ml (100 μl/well) can bind Madarex LAG-3 Mab, Human IgG1 with a linear range of 20-78 ng/ml (QC tested).
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    FDC|CD223|LAG3
  • Protein/peptide name:
    LAG-3
  • Purity:
    >90%
  • Molecular weight:
    73,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7,5. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

About this item