Human recombinant complement factor D (from HEK293 cells)
Marketsource
Supplier: ACRO Biosystems
Human recombinant complement factor D (from HEK293 cells)
Supplier: ACRO Biosystems
Marketsource
Certificates
About this item
Human Complement Factor D / CFD Protein, His Tag (active enzyme), ACROBiosystems.
2 Options Available Below
Product Details & Documents
Human Complement Factor D / CFD Protein, His Tag (active enzyme), ACROBiosystems.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ACRBCFD-H52H8-50UG
- ACRBCFD-H52H8-1MG
- Protein/peptide name:
- Complement Factor D
- Complement Factor D
- Protein synonyms:
- PFD|Complement factor D|CFD|DF|Adipsin
- PFD|Complement factor D|CFD|DF|Adipsin
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Source:
- HEK293 cells
- HEK293 cells
- Conjugation:
- Unconjugated
- Unconjugated
- Gene ID:
- P00746-1
- P00746-1
- Endotoxin level:
- Low
- Low
- Endotoxin content:
- Less than 1,0 EU per μg by the LAL method.
- Less than 1,0 EU per μg by the LAL method.
- Purity:
- >90% as determined by SDS-PAGE.
- >90% as determined by SDS-PAGE.
- Formulation:
- Lyophilised from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH8.0. Normally glycerol is added as protectant before Lyophilisation.
- Lyophilised from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH8.0. Normally glycerol is added as protectant before Lyophilisation.
- Molecular weight:
- 26,3 kDa
- 26,3 kDa
- Storage conditions:
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- Tag sequence:
- His Tag (active enzyme)
- His Tag (active enzyme)
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.ACRBCFD-H52H8-50UGProtein/peptide nameComplement Factor DProtein synonymsPFD|Complement factor D|CFD|DF|AdipsinProtein/peptide typeRecombinantSpeciesHumanSourceHEK293 cellsConjugationUnconjugatedGene IDP00746-1Endotoxin levelLowEndotoxin contentLess than 1,0 EU per μg by the LAL method.Purity>90% as determined by SDS-PAGE.FormulationLyophilised from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH8.0. Normally glycerol is added as protectant before Lyophilisation.Molecular weight26,3 kDaStorage conditions–20 °CShipping temperatureRTTag sequenceHis Tag (active enzyme)Marketsource - Specifications:Marketsource
More
Specifications
Cat. No.ACRBCFD-H52H8-1MGProtein/peptide nameComplement Factor DProtein synonymsPFD|Complement factor D|CFD|DF|AdipsinProtein/peptide typeRecombinantSpeciesHumanSourceHEK293 cellsConjugationUnconjugatedGene IDP00746-1Endotoxin levelLowEndotoxin contentLess than 1,0 EU per μg by the LAL method.Purity>90% as determined by SDS-PAGE.FormulationLyophilised from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH8.0. Normally glycerol is added as protectant before Lyophilisation.Molecular weight26,3 kDaStorage conditions–20 °CShipping temperatureRTTag sequenceHis Tag (active enzyme)Marketsource



