Human recombinant FAP (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
FITC-Labeled Human FAP Protein, His Tag (MALS verified), ACROBiosystems.
Specifications
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag (MALS verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:2e5 of anti-FAP CAR 293 cells were stained with 100 μL of 3 μg/ml of FITC-Labeled Human FAP, His Tag (Cat. No. FAP-HF243) and negative control protein respectively, FITC signals was used to evaluate the binding activity (QC tested).
- Gene ID:Q12884-1
- Protein synonyms:FAP|APCE|SIMP|Seprase|FAPalpha
- Protein/peptide name:FAP
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:86,9 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before Lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together