Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant FOLR1 (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Human recombinant FOLR1 (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Supplier:  ACRO Biosystems
Certificates
Human recombinant FOLR1 (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag (MALS verified)
  • Storage conditions:
    –20 ℃, aviod light
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    2e5 of Anti-FOLR1 CAR-293 cells were stained with 100 μL of 3 μg/ml of FITC-Labeled Human FOLR1, His Tag (Cat. No. FO1-HF2H8) and negative control protein respectively, FITC signals was used to evaluate the binding activity (QC tested).
  • Gene ID:
    P15328-1
  • Protein synonyms:
    FOLR-1|FOLR|FBP
  • Protein/peptide name:
    FOLR1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    26,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before Lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together