Cynomolgus recombinant Fc gamma RIII / CD16 (from HEK293)
Catalog # ACRBFC6-C52H9-1MG
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Pack type:1 mg
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Rituximab at 5 μg/ml (100 μl/well) can bind Cynomolgus CD16, His Tag (BLI verified) with a linear range of 39-313 ng/ml (QC tested); Captured Cynomolgus CD16, His Tag (BLI verified)on CM5 chip via anti-His antibody can bind MabThera® (Rituximab) with an affinity constant of 0,206 μM as determined in a SPR assay (Biacore T200) (Routinely tested).; Loaded Cynomolgus CD16, His Tag (BLI verified) on HIS1K Biosensor, can bind Rituximab with an affinity constant of 0,287 μM as determined in BLI assay (ForteBio Octet Red96e) (QC tested).; Loaded Herceptin on Protein A Biosensor, can bind Cynomolgus CD16, His Tag (BLI verified) with an affinity constant of 0,31 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Herceptin on FAB2G Biosensor, can bind Cynomolgus CD16, His Tag (BLI verified) with an affinity constant of 0,23 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested)
- Gene ID:Q8SPW2-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:FCGR3
- Protein/peptide name:Fc gamma RIII / CD16
- Purity:>95%
- Molecular weight:23,8 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder,RT