Cynomolgus recombinant DNAM-1 (from HEK293)
Catalog # ACRBDN1-C52H9-1MG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pack type:1 mg
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Cynomolgus DNAM-1, His Tag at 2 μg/ml (100 μl/well) can bind Human CD155, Mouse IgG2a Fc Tag, low endotoxin with a linear range of 2-40 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Cynomolgus DNAM-1, His Tag at 2 μg/ml (100 μl/well) can bind Human Nectin-2, Fc Tag with a linear range of 5-40 ng/ml (Routinely tested).; Loaded Human CD155, Fc Tag on Protein A Biosensor, can bind Cynomolgus DNAM-1, His Tag with an affinity constant of 0,88 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested)
- Gene ID:A0A2K5W1Z6-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD226|PTA1|DNAM1
- Protein/peptide name:DNAM-1
- Purity:>90%
- Molecular weight:28,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder,RT