Human recombinant TrkA (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human TrkA / NTRK1 (33-417) Protein, Mouse IgG2a Fc Tag.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Mouse IgG2a Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method
- Gene ID:P04629-2
- Protein synonyms:TRKA|Trk-A|MTC|NTRK1|TrkA|TRK
- Protein/peptide name:TrkA
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:68,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together