Human recombinant Serum Albumin (from HEK293 cells), Biotin
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Biotinylated Human Serum Albumin Protein, His,Avitag™.
Specifications
- Conjugation:Biotin
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His, Avitag™
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Loaded Biotinylated Human Serum Albumin, His,Avitag (Cat. No. HSA-H82E3) on SA Biosensor, can bind Human FCGRT&B2M Heterodimer Protein, His Tag&Strep II Tag (SPR & BLI verified) (Cat. No. FCM-H5286) with an affinity constant of 0,641 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P02768-1
- Protein synonyms:Serum albumin|Alb|ALB
- Protein/peptide name:Serum Albumin
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:70,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together