Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant PSMA (from HEK293 cells)
Human recombinant PSMA (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Human recombinant PSMA (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    FACS analysis of Human PSMA, Fc Tag binding to PSMA-CAR-293 cells transfected with anti-PSMA-scFv 2X105 of PSMA-CAR-293 cells transfected with anti-PSMA-scFv were stained with 100μl of 1μg/ml of Human PSMA, Fc Tag (Cat. No. PSA-H5264) and negative control protein respectively, washed and then followed by PE anti-human IgG Fc antibody and analysed with FACS (Routinely tested).
  • Gene ID:
    Q04609-1 
  • Protein synonyms:
    FOLH|FOLH1|PSM|GCPII|NAALAD1|PSMA|FGCP|NAALADase I|GIG27
  • Protein/peptide name:
    PSMA
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    106,0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together