Human recombinant PLAU (from HEK293 cells)
Human PLAU / uPA Protein, His Tag (activated by trypsin) (active enzyme).
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag (activated by trypsin) (active enzyme)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Gene ID:P00749-1
- Protein synonyms:URK|ATF|u-PA|Urokinase|PLAU|QPD|UPA|BDPLT5
- Protein/peptide name:PLAU
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:15,3 kDa, 17,8 kDa and 29,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 25 mM HEPES, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Frequently Bought Together