Human recombinant GITR (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human GITR, Fc Tag (ACRBGIR-H5254-100) at 2 μg/ml (100 μl/well) can bind Biotinylated Human GITR Ligand, Avitag,Fc Tag (ACRBGIR-H5254-100) with a linear range of 4 - 63 ng/ml (QC tested).
- Gene ID:Q9Y5U5-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD357|GITR|TNFRSF18|AITR
- Protein/peptide name:GITR
- Purity:>95%
- Molecular weight:41,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder, RT
Specifications
Frequently Bought Together