Human recombinant Fc gamma RIIIB / CD16b (NA2) (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation. Biotinylated Human CD16b (NA2), His,Avitag (SPR and BLI verified) (ACRBCDB-H82EA-200) captured on Biotin CAP- Series S Sensor Chip can bind Rituximab with an affinity constant of 4,22 μM as determined in a SPR assay (Biacore T200) (QC tested). Immobilised Biotinylated Human CD16b (NA2), His,Avitag (SPR and BLI verified) (ACRBCDB-H82EA-200) on SA Chip can bind Rituximab with an affinity constant of 6,66 μM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Biotinylated Human CD16b (NA2), His,Avitag (SPR and BLI verified)(ACRBCDB-H82EA-200) on SA Biosensor, can bind Rituximab with an affinity constant of 5,3 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:O75015
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:FCGR3B|Fc gamma RIIIB|FCGR3|FCG3B|FCG3|CD16B|IGFR3|CD16b (NA2)
- Protein/peptide name:Fc gamma RIIIB / CD16b (NA2)
- Purity:>95%
- Molecular weight:24,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder, RT
Specifications
Frequently Bought Together