Order Entry
Switzerland
ContactUsLinkComponent
 
Human recombinant Fc gamma RIIIB / CD16b (NA2) (from HEK293 cells)
Catalog # ACRBCDB-H5222-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Human recombinant Fc gamma RIIIB / CD16b (NA2) (from HEK293 cells)
Catalog # ACRBCDB-H5222-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Seq:
    02
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation. Immobilised Human CD16b (NA2) (ACRBCDB-H5222-1MG) on CM5 Chip via anti-His antibody, can bind Rituximab with an affinity constant of 2,88 μM as determined in a SPR assay (Biacore T200) (QC tested).
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    FCGR3B|Fc gamma RIIIB|FCGR3|FCG3B|FCG3|CD16B|IGFR3|CD16b (NA2)
  • UniProtKB:
    O75015-1
  • Protein/peptide name:
    Fc gamma RIIIB / CD16b (NA2)
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    22,7 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    Powder, RT

Specifications

About this item