Human recombinant CD7 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Llama IgG2b Fc Tag
- Storage conditions:−20 °C
- Endotoxin content:Less than 0,01 EU per μg by the LAL method
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised human CD7, Llama IgG2b Fc Tag, low endotoxin (Cat. No. ACRBCD7-H5258) at 5 μg/ml (100 μl/well) can bind Human SECTM1, His Tag (Cat. No. ACRBSE1-H5227) with a linear range of 10 - 39 ng/ml (QC tested).
- Gene ID:P09564-1
- Protein synonyms:LEU-9|TP41|CD7|Tp40|GP40
- Protein/peptide name:CD7
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:44,3 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with glycine, arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together