Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant VEGF R3 (from HEK293)
Mouse recombinant VEGF R3 (from HEK293)
Catalog # ACRBFL4-M5251100UG
Supplier:  ACRO Biosystems
CAS Number:  
Mouse recombinant VEGF R3 (from HEK293)
Catalog # ACRBFL4-M5251100UG
Supplier:  ACRO Biosystems
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Less than 0,1 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised human VEGF-C, His Tag (Cat. No. ACRBVEC-H4225) at 5 μg/ml (100 μl/well) can bind Mouse VEGF R3, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBFL4-M5251) with a linear range of 1 - 10 ng/ml (QC tested).
  • Gene ID:
    P35917-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    Flt-4|VEGFR3|FLT4|LMPH1A|VEGFR-3|PCLFLT41|FLT-41|FLT41|FLT-4|PCL
  • Protein/peptide name:
    VEGF R3
  • Purity:
    >95%
  • Molecular weight:
    111,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

About this item