Mouse recombinant PD-L2 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with glycine, arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised mouse PD-L2, Fc Tag (Cat. No. ACRBPD2-M5254) at 5 μg/ml (100 μl/well) can bind mouse PD-1, mouse IgG2a Fc tag (Cat. No. ACRBPD1-M52A4) with a linear range of 0,078 - 0,625 μg/ml (QC tested).
- Gene ID:Q9WUL5
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:PDCD1L2|CD273|PD-L2|Butyrophilin B7-DC|PDL2|PDCD1LG2|PDCD1 ligand 2
- Protein/peptide name:PD-L2
- Purity:>95%
- Molecular weight:49,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with glycine, arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together