Mouse recombinant IL-17A (from HEK293)
Catalog # ACRBCT8-M5240100UG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pk:100 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Loaded Human IL-17 RA, Fc Tag (Cat. No. ACRBILA-H5257) on protein A biosensor, can bind mouse IL-17A, His tag (Cat. No. ACRBCT8-M5240) with an affinity constant of 20,1 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Mouse IL-17A, His Tag (Cat. No. ACRBCT8-M5240) stimulates secretion of IL-6 by NIH-3T3 cells. The ED50 for this effect is 2,132 - 5,005 ng/ml.
- Gene ID:Q62386-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:IL-17|IL-17A|CTLA-8|Interleukin-17A
- Protein/peptide name:IL-17A
- Purity:>90%
- Molecular weight:15,8 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT