Human recombinant IL-13 R alpha 2 (from HEK293 cells), FITC (fluorescein isothiocyanate)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage conditions:–20 °C, avoid light
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised Human IL-13, His Tag (Cat. No. IL3-H52H4) at 5 μg/ml (100 μl/well) can bind FITC-Labeled Human IL-13 R alpha 2, His Tag (Cat. No. IL2-HF2H3) with a linear range of 0,313 - 1,25 μg/ml (QC tested).
- Gene ID:Q14627-1
- Protein synonyms:IL13RA2|CD213A2|IL-13R|CT19|IL13BP
- Protein/peptide name:IL-13 R alpha 2
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:39,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together