Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant IL-12 R beta 1 (from HEK293 cells)
Human recombinant IL-12 R beta 1 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant IL-12 R beta 1 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilised Human IL-12 R beta 1, His Tag (Cat. No. ILB-H52H9) at 2 μg/ml (100 μl/well) can bind Human IL-12B, Fc Tag (Cat. No. NK2-H5258) with a linear range of 0,078 -1,25 μg/ml (QC tested).
  • Gene ID:
    P42701-1
  • Protein synonyms:
    CD212|IL12RB|IL-12 R beta 1|IL12RB1|IL-12RB1|IL12R
  • Protein/peptide name:
    IL-12 R beta 1
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    59,0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together