Human recombinant IFN-alpha 1 (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:P01562-1
- Protein synonyms:LeIF D|IFNA1|IFN-alpha-1|IFN-alpha 1|IFNalpha 1
- Protein/peptide name:IFN-alpha 1
- Molecular weight:45,8 kDa
- Endotoxin level:Low
- Formulation:Please contact us for detailed information.
- Shipping temperature:RT
Specifications
Frequently Bought Together